Tris(dibenzylideneacetone)dipalladium(0)

Supplier: APOLLO SCIENTIFIC
Warning

IN2823-1G IN2823-25G IN2823-5G
76615-092EA 30.56 USD
76615-092 76615-094 76615-096
Tris(dibenzylideneacetone)dipalladium(0)
Tris(dibenzylideneacetone)dipalladium(0)
Formula: C₅₁H₄₂O₃Pd₂
Melting Pt: 152…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013310
CAS Number: 51364-51-3
EINECS: 479-280-5
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR