tert-Butyl-3-iodoazetidine-1-carboxylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tert-butyl 3-iodoazetidine-1-carboxylate, 1-BOC-3-iodoazetidine, tert-Butyl 3-iodoazetidine-1-carboxylate, N-Boc-3-iodoazetidine

OR2251-100G OR2251-25G OR2251-5G
76616-052EA 140.58 USD
76616-052 76616-054 76616-056
tert-Butyl-3-iodoazetidine-1-carboxylate 97%
tert-Butyl-3-iodoazetidine-1-carboxylate
Formula: C₈H₁₄INO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09752821
CAS Number: 254454-54-1
UN: 3082
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR