4-Bromo-7-azaindole

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-7-azaindole, 4-Bromo-1H-pyrrolo[2,3-b]pyridine

OR42186-1G OR42186-25G OR42186-5G
76616-718EA 20.97 USD
76616-718 76616-720 76616-722
4-Bromo-7-azaindole
4-Bromo-7-azaindole
Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 176…183 °C
Storage Temperature: Ambient
MDL Number: MFCD08272233
CAS Number: 348640-06-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR