5-Acetyl-2-chloropyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Acetyl-6-chloropyridine

OR25927-1G OR25927-5G
76616-170EA 20.34 USD
76616-170 76616-172
5-Acetyl-2-chloropyridine
5-Acetyl-2-chloropyridine
Formula: C₇H₆ClNO
MW: 155.5816 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD03407343
CAS Number: 55676-22-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR