1-Ethoxy-1-(tributylstannyl)ethene 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Tri-n-butyl(1-ethoxyvinyl)tin
, 1-Ethoxyvinyltri-n-butyltin
, Tributyl(1-ethoxyvinyl)tin
OR2395-25G
OR2395-100G
OR2395-5G
76611-388EA
124
USD
76611-388
76611-390
76611-392
1-Ethoxy-1-(tributylstannyl)ethene 95%
1-Ethoxy-1-(tributylstannyl)ethene
Formula:
C₁₆H₃₄OSn MW: 361.15 g/mol Boiling Pt: 85…86 °C/0.1mmHg Density: 1.069 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010240 CAS Number: 97674-02-7 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...