5-Allyl-3-methoxysalicylaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Allyl-2-hydroxy-3-methoxybenzaldehyde

OR55035-1G OR55035-5G
76592-198EA 48.9 USD
76592-198 76592-200
5-Allyl-3-methoxysalicylaldehyde
5-Allyl-3-methoxysalicylaldehyde
Formula: C₁₁H₁₂O₃
Boiling Pt: 167 °C/9.75mmHg
Melting Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD06408022
CAS Number: 22934-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR