3-Ethynyltoluene

Supplier: APOLLO SCIENTIFIC
Warning

OR3481-1G OR3481-25G OR3481-5G
76625-146EA 20.28 USD
76625-146 76625-148 76625-150
3-Ethynyltoluene
3-Ethynyltoluene
Formula: C₉H₈
Boiling Pt: 170…175 °C
Density: 0.9 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD01318174
CAS Number: 766-82-5
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR