2-Methoxy-6-(tributylstannyl)pyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR15611-1G OR15611-500MG
76611-176EA 99.82 USD
76611-176 76611-178
2-Methoxy-6-(tributylstannyl)pyridine
2-Methoxy-6-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NOSn
Storage Temperature: Refrigerator
MDL Number: MFCD09025803
CAS Number: 164014-94-2
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR