2-Amino-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: [1,3,4]Thiadiazol-2-ylamine, 1,3,4-Thiadiazol-2-amine

OR23108-100G OR23108-25G OR23108-5G
76616-088EA 29.23 USD
76616-088 76616-090 76616-092
2-Amino-1,3,4-thiadiazole
2-Amino-1,3,4-thiadiazole
Formula: C₂H₃N₃S
MW: 101.13
Melting Pt: 188…194 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003107
CAS Number: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR