1,8-Diazabicyclo[5.4.0]undec-7-ene
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
DBU
, 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine
OR1967-1KG
OR1967-500G
OR1967-250G
76624-872EA
85
USD
76624-872
76624-874
76624-876
1,8-Diazabicyclo[5.4.0]undec-7-ene
1,8-Diazabicyclo[5.4.0]undec-7-ene
Formula:
C₉H₁₆N₂ MW: 152.24 g/mol Boiling Pt: 80…83 °C Melting Pt: –70 °C Density: 1.018 g/cm³ (25 °C) Flash Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006930 CAS Number: 6674-22-2 EINECS: 229-713-7 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...