Cyclohexanecarbonitrile

Supplier: APOLLO SCIENTIFIC
Danger

OR27707-1G OR27707-5G
76625-008EA 20.26 USD
76625-008 76625-010 76625-012
Cyclohexanecarbonitrile
Cyclohexanecarbonitrile
Formula: C₇H₁₁N
MW: 109.17 g/mol
Boiling Pt: 75…76 °C/16mmHg
Melting Pt: 11 °C
Density: 0.919 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00040444
CAS Number: 766-05-2
EINECS: 212-157-4
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR