4-Picolyl chloride hydrochloride

Supplier: APOLLO SCIENTIFIC
Danger

OR0263-100G OR0263-25G OR0263-500G
76615-220EA 55.02 USD
76615-220 76615-222 76615-224
4-Picolyl chloride hydrochloride
4-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
Melting Pt: 166…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012826
CAS Number: 1822-51-1
EINECS: 217-350-7
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR