Benzoin methyl ether 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone
OR11336-100G
OR11336-25G
76615-464EA
110.02
USD
76615-464
76615-466
Benzoin methyl ether 98%
Benzoin methyl ether
Formula:
C₁₅H₁₄O₂ Melting Pt: 47…50 °C Density: 1.128 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008492 CAS Number: 3524-62-7 EINECS: 222-538-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...