3-Bromoanthranilic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Amino-3-bromobenzoic acid

OR14301-100G OR14301-25G OR14301-5G
76615-696EA 139.56 USD
76615-696 76615-698 76615-700
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
Boiling Pt: 343…344 °C
Melting Pt: 174…178 °C
Density: 1.793 g/cm³ (25 °C)
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD03618453
CAS Number: 20776-51-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR