Methoxyacetyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR40393-100G OR40393-25G OR40393-10G
76616-690EA 108 USD
76616-690 76616-692 76616-694 77409-534
Methoxyacetyl chloride 95%
Methoxyacetyl chloride
Formula: C₃H₅ClO₂
Boiling Pt: 112…113 °C
Density: 1.187 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000728
CAS Number: 38870-89-2
EINECS: 254-169-2
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR