1,10-Phenanthroline hydrochloride monohydrate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate

OR2360-100G OR2360-250G
76616-114EA 143.64 USD
76616-114 76616-116
1,10-Phenanthroline hydrochloride monohydrate 97%
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
Melting Pt: 224…225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR