tert-Butyl cyanoacetate

Supplier: APOLLO SCIENTIFIC
Warning

OR55135-100G OR55135-250G OR55135-25G
76592-272EA 74.38 USD
76592-272 76592-274 76592-270
tert-Butyl cyanoacetate
tert-Butyl cyanoacetate
Formula: C₇H₁₁NO₂
Boiling Pt: 67…68 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001938
CAS Number: 1116-98-9
EINECS: 214-243-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR