Maleimide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1H-Pyrrole-2,5-dione

OR5487-100G OR5487-25G
76616-992EA 94.74 USD
76616-992 76616-994
Maleimide
Maleimide
Formula: C₄H₃NO₂
MW: 97.07 g/mol
Boiling Pt: 97…103 °C/5mmHg
Melting Pt: 91…93 °C
Density: 1.249 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
EINECS: 208-787-4
UN: 2923
ADR: 8+6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR