4-Bromo-o-phenylenediamine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Bromo-3,4-diaminobenzene, 4-Bromobenzene-1,2-diamine, 4-Bromo-1,2-phenylenediamine, 4-Bromo-1,2-diaminobenzene, 2-Amino-4-bromophenylamine

OR22977-100G OR22977-25G OR22977-5G
76616-062EA 109 USD
76616-062 76616-064 76616-066
4-Bromo-o-phenylenediamine 99%
4-Bromo-o-phenylenediamine
Formula: C₆H₇BrN₂
MW: 187.04
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD02660622
CAS Number: 1575-37-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR