Potassium tetrachloroplatinate(II) 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Dipotassium tetrachloroplatinate, Potassium tetrachloroplatinate

IN1004-1G IN1004-5G
76624-620EA 69.26 USD
76624-620 76624-622
Potassium tetrachloroplatinate(II) 95%
Potassium tetrachloroplatinate (II)
Formula: Cl₄K₂Pt
Density: 3.38 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR