1,4-Diazabicyclo[2.2.2]octane

Supplier: APOLLO SCIENTIFIC
Danger

OR322352-100G OR322352-1KG OR322352-500G
76611-704EA 20.34 USD
76611-704 76611-706 76611-708
1,4-Diazabicyclo[2.2.2]octane
1,4-Diazabicyclo[2.2.2]octane
Formula: C₆H₁₂N₂
Boiling Pt: 174 °C/760mmHg
Melting Pt: 156…159 °C
Density: 1.02 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006689
CAS Number: 280-57-9
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR