1-Bromo-4-ethynylbenzene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-Bromophenylacetylene, 1-Ethynyl-4-bromobenzene, (4-Bromophenyl)acetylene, (4-Bromophenyl)ethyne
OR40459-25G
OR40459-5G
76616-702EA
433.63
USD
76616-702
76616-704
76616-706
1-Bromo-4-ethynylbenzene 98%
1-Bromo-4-ethynylbenzene
Formula:
C₈H₅Br MW: 181.03 g/mol Boiling Pt: 88…90 °C/16mmHg Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00168822 CAS Number: 766-96-1 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...