4-Nitrobenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Nitrobenzene-1-sulfonyl chloride, 4-Nitrobenzene-1-sulphonyl chloride

OR28919-100G OR28919-25G OR28919-5G
76620-962EA 116.74 USD
76620-962 76620-964 76620-966
4-Nitrobenzenesulfonyl chloride
4-Nitrobenzenesulphonyl chloride
Formula: C₆H₄ClNO₄S
MW: 221.62
Boiling Pt: 350
Melting Pt: 79
Density: 1.69
Storage Temperature: Ambient
MDL Number: MFCD00007445
CAS Number: 98-74-8
EINECS: 202-697-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR