4-Chlorobenzenesulfonyl chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR4476-500G OR4476-100G OR4476-25G
76621-400EA 167.06 USD
76621-400 76621-396 76621-398
4-Chlorobenzenesulfonyl chloride 99%
4-Chlorobenzenesulphonyl chloride
Formula: C₆H₄Cl₂O₂S
Boiling Pt: 141 °C/15mmHg
Melting Pt: 50…52 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00007439
CAS Number: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR