3,5-Dichlorobenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR5384-100G OR5384-25G OR5384-5G
76621-616EA 117.16 USD
76621-616 76621-618 76621-620
3,5-Dichlorobenzenesulfonyl chloride
3,5-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
Boiling Pt: 85…86 °C/0.5mmHg
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR