Trichloroacetonitrile 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR4786-1KG OR4786-250G OR4786-5KG
76625-232EA 166.04 USD
76625-232 76625-234 76625-236
Trichloroacetonitrile 99%
Trichloroacetonitrile
Formula: C₂Cl₃N
MW: 144.39 g/mol
Boiling Pt: 85.7
Melting Pt: -44
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001842
CAS Number: 545-06-2
EINECS: 208-885-7
UN: 3276
ADR: 6.1 III
Merck Index: 12,09757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR