Pyrrolidine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Tetrahydropyrrole

OR0971-100ML
76610-882EA 16 USD
76610-882
Pyrrolidine 99%
Pyrrolidine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 86…88 °C
Melting Pt: –60 °C
Density: 0.856 g/cm³ (25 °C)
Flash Pt: 3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005249
CAS Number: 123-75-1
EINECS: 204-648-7
UN: 1922
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR