α,2,6-Trichlorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,6-Dichlorobenzyl Chloride1,3-Dichloro-2-(chloromethyl)benzene2,6-Dichlorobenzyl chloride1-(Chloromethyl)-2,6-dichlorobenzene

OR28624-1KG OR28624-250G
76620-938EA 21.63 USD
76620-938 76620-936
α,2,6-Trichlorotoluene 97%
α,2,6-Trichlorotoluene
Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 117…119 °C/14mmHg
Melting Pt: 36…39 °C
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000897
CAS Number: 2014-83-7
EINECS: 217-940-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR