3-Aminophenol
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
m-Aminophenol
, 3-Hydroxyaniline
OR16952-100G
OR16952-1KG
OR16952-500G
76611-224EA
21
USD
76611-224
76611-226
76611-228
3-Aminophenol
3-Aminophenol
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 164 °C/11mmHg Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007786 CAS Number: 591-27-5 EINECS: 209-711-2 UN: 2512 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...