1-Methylcyclopropanecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-methylcyclopropane-1-carboxylic acid

OR24738-10G OR24738-1G OR24738-5G
76620-852EA 85.58 USD
76620-852 76620-854 76620-856
1-Methylcyclopropanecarboxylic acid
1-Methylcyclopropanecarboxylic acid
Formula: C₅H₈O₂
Boiling Pt: 183…185 °C
Melting Pt: 30 °C
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00001290
CAS Number: 6914-76-7
EINECS: 230-020-7
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR