Hydrobromic acid 48% in aqueous solution

Supplier: APOLLO SCIENTIFIC
Danger

OR13036-1KG OR13036-5KG
76611-072EA 36.66 USD
76611-072 76611-074
Hydrobromic acid 48% in aqueous solution
Hydrogen bromide
Formula: HBr
Boiling Pt: 126…128 °C
Melting Pt: –11 °C
Density: 1.48 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR