Imidazole

Supplier: APOLLO SCIENTIFIC
Danger

OR5102-1KG OR5102-2.5KG OR5102-500G
76621-520EA 38.72 USD
76621-520 76621-522 76621-524
Imidazole
Imidazole
Formula: C₃H₄N₂
Boiling Pt: 235 °C
Melting Pt: 85…91 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR