Diphenyl ether

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

OR46509-100G OR46509-1KG OR46509-500G
76616-840EA 20.87 USD
76616-840 76616-842 76616-844
Diphenyl ether
Diphenyl ether
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 498
Melting Pt: 25…27 °C
Density: 1.073 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: 9+EHS III
Merck Index: 12,07442

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR