Maleic hydrazide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,2-Dihydro-3,6-pyridazinedione, 3,6-Dihydroxypyridazine, 3,6-Pyridazinediol, Maleic acid Hydrazide

OR4916-5G OR4916-100G OR4916-25G
76611-948EA 20.99 USD
76611-948 76611-944 76611-946
Maleic hydrazide
Maleic hydrazide
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 292 (dec.)
Density: 1.6 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006665
CAS Number: 123-33-1
EINECS: 204-619-9
UN: 1325
ADR: 4.1 III
Merck Index: 13,05728

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR