2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: DDQ, 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-Dichloro-5,6-dicyanoquinone, 4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile

OR14321-1KG OR14321-250G OR14321-500G
76624-786EA 370.8 USD
76624-786 76624-788 76624-790
2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%
2,3-Dichloro-5,6-dicyano-p-benzoquinone
Formula: C₈Cl₂N₂O₂
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00001593
CAS Number: 84-58-2
EINECS: 201-542-2
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR