α,α'-Dibromo-m-xylene 97+%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR26233-25G
OR26233-100G
OR26233-10G
76620-900EA
38.72
USD
76620-900
76620-896
76620-898
α,α'-Dibromo-m-xylene 97+%
α,α'-Dibromo-m-xylene
Formula:
C₈H₈Br₂ Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000178 CAS Number: 626-15-3 EINECS: 210-931-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...