α-Bromo-3-chlorotoulene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-m-chlorotoulene, 3-Chlorobenzyl bromide, 1-(Bromomethyl)-3-chlorobenzene, alpha-Bromo-3-chlorotoluene

OR5035-100G OR5035-25G
76621-498EA 78.44 USD
76621-498 76621-500
α-Bromo-3-chlorotoulene 98%
α-Bromo-3-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48
Boiling Pt: 238
Melting Pt: 17.5
Density: 1.569 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS Number: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR