4-Nitrophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol

OR322090-100G OR322090-25G OR322090-5G
76611-688EA 46.86 USD
76611-688 76611-690 76611-692
4-Nitrophenol 98%
4-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 279 °C
Melting Pt: 114 °C
Density: 1.48 g/cm³ (25 °C)
Flash Pt: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD00007331
CAS Number: 100-02-7
UN: 1663
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR