trans-1,4-Dibromo-2-butene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene

OR3193-100G OR3193-25G OR3193-500G
76616-448EA 39.4 USD
76616-448 76616-450 76616-452
trans-1,4-Dibromo-2-butene
trans-1,4-Dibromo-2-butene
Formula: C₄H₆Br₂
MW: 213.9
Boiling Pt: 203
Melting Pt: 48…53 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000249
CAS Number: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8+6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR