Bromotrichloromethane
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR43546-25G
OR43546-100G
76611-848EA
34.64
USD
76611-848
76611-846
Bromotrichloromethane
Bromotrichloromethane
Formula:
BrCl₃C Boiling Pt: 104…105 °C Melting Pt: –6 °C Density: 2.015 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000783 CAS Number: 75-62-7 EINECS: 200-886-0 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...