Potassium carbonate, anhydrous 99%
Supplier: APOLLO SCIENTIFIC
Synonyms:
Carbonic acid potassium salt
, Potash
, Carbonic acid, dipotassium salt
IN14572A-100G
IN14572A-1KG
76563-020EA
39.14
USD
76563-020
76560-152
Potassium carbonate, anhydrous 99%
Potassium carbonate
Formula:
K₂CO₃ MW: 138.21 g/mol Melting Pt: 891 °C Density: 2.43 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011382 CAS Number: 584-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...