2'-sulfamoyl-[1,1'-biphenyl]-4-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(2-sulfamoylphenyl)benzoic acid, 4-(2-Sulphamoylphenyl)benzoic acid, 2'-(Aminosulphonyl)-[1,1'-biphenyl]-4-carboxylic acid, 2'-(Aminosulfonyl)-[1,1'-biphenyl]-4-carboxylic acid

OR6989-1G OR6989-250MG
76587-328EA 575.1 USD
76587-328 76587-326
2'-sulfamoyl-[1,1'-biphenyl]-4-carboxylic acid
2'-sulfamoyl-[1,1'-biphenyl]-4-carboxylic acid
Formula: C₁₃H₁₁NO₄S
Storage Temperature: Ambient
MDL Number: MFCD03001764
CAS Number: 352615-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR