2,5,6,7-Tetrahydro-4H-indazol-4-one 97
Supplier: APOLLO SCIENTIFIC
OR510001-1G
OR510001-250MG
76586-384EA
263.84
USD
76586-384
76586-382
2,5,6,7-Tetrahydro-4H-indazol-4-one 97
2,5,6,7-Tetrahydro-4H-indazol-4-one
Formula:
C₇H₈N₂O MW: 136.151 g/mol Storage Temperature: Refrigerator |
CAS Number:
912259-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...