3-Bromo-2-nitrophenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR50991-1G OR50991-250MG
76586-374EA 681.48 USD
76586-374 76586-372
3-Bromo-2-nitrophenylacetic acid
3-Bromo-2-nitrophenylacetic acid
Formula: C₈H₆BrNO₄
Storage Temperature: Ambient
MDL Number: MFCD18392318
CAS Number: 400629-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR