Ethyl 5-bromothiophene-3-carboxylate 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR46588-1G OR46588-5G
76585-130EA 20.34 USD
76585-130 76585-132
Ethyl 5-bromothiophene-3-carboxylate 97+%
Ethyl 5-bromothiophene-3-carboxylate
Formula: C₇H₇BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD14155650
CAS Number: 170355-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR