Ethyl 6-chlorobenzofuran-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR45160-1G OR45160-250MG
76584-566EA 835.06 USD
76584-566 76584-564
Ethyl 6-chlorobenzofuran-2-carboxylate
Ethyl 6-chlorobenzofuran-2-carboxylate
Formula: C₁₁H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD28987315
CAS Number: 74222-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR