4-Bromo-5-methylthiophene-2-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR451423-1G OR451423-5G
76584-430EA 72.32 USD
76584-430 76584-432
4-Bromo-5-methylthiophene-2-carbaldehyde
4-Bromo-5-methylthiophene-2-carbaldehyde
Formula: C₆H₅BrOS
Storage Temperature: Refrigerator
MDL Number: MFCD01118659
CAS Number: 29421-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR