2-Bromo-4-iodo-6-methylanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-5-iodo-2-methoxy-3-methylbenzene, 2-Bromo-4-iodo-6-methylphenyl methyl ether, 3-Bromo-5-iodo-2-methoxytoluene

OR45135-1G OR45135-250MG OR45135-5G
76584-364EA 220.04 USD
76584-364 76584-362 76584-366
2-Bromo-4-iodo-6-methylanisole
2-Bromo-4-iodo-6-methylanisole
Formula: C₈H₈BrIO
Storage Temperature: Ambient
MDL Number: MFCD27937157
CAS Number: 1820675-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR