5-Bromothiazolidine-2,4-dione

Supplier: APOLLO SCIENTIFIC

OR450092-1G OR450092-250MG
76584-096EA 756.86 USD
76584-096 76584-094
5-Bromothiazolidine-2,4-dione
5-Bromothiazolidine-2,4-dione
Formula: C₃H₂BrNO₂S
MW: 196.022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03305737
CAS Number: 125518-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR