2-Iodo-6-methoxyaniline

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Iodo-o-anisidine, 2-Amino-3-iodoanisole

OR43627-1G OR43627-250MG
76583-904EA 62.14 USD
76583-904 76583-902 76583-906
2-Iodo-6-methoxyaniline
2-Iodo-6-methoxyaniline
Formula: C₇H₈INO
Storage Temperature: Ambient
MDL Number: MFCD11977372
CAS Number: 354574-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR